3-Brom-2-fluor-5-methylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-2-fluoro-5-picoline
AS50411G1
AS50411G5
ACTIAS50411G1EA
0
CHF
ACTIAS50411G1
ACTIAS50411G5
3-Brom-2-fluor-5-methylpyridin
3-Bromo-2-fluoro-5-methylpyridine
Formel:
C₆H₅BrFN MW: 190.01 g/mol |
MDL Number:
MFCD03095305 CAS-Nummer: 17282-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...