2-Brom-3-fluor-6-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-3-fluoro-6-picoline

AS31827G1 AS31827G5
ACTIAS31827G1EA 0 CHF
ACTIAS31827G1 ACTIAS31827G5
2-Brom-3-fluor-6-methylpyridin
2-Bromo-3-fluoro-6-methylpyridine
Formel: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03095061
CAS-Nummer: 374633-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR