2-Brom-3-fluor-4-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-3-fluoro-4-picoline

AS34987G1 AS34987G10 AS34987G5
ACTIAS34987G1EA 0 CHF
ACTIAS34987G1 ACTIAS34987G10 ACTIAS34987G5
2-Brom-3-fluor-4-methylpyridin
2-Bromo-3-fluoro-4-methylpyridine
Formel: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD04112499
CAS-Nummer: 884494-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR