2-Brom-4-nitrobenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS33717G1
AS33717G10
AS33717G5
ACTIAS33717G1EA
0
CHF
ACTIAS33717G1
ACTIAS33717G10
ACTIAS33717G5
2-Brom-4-nitrobenzoesäure
2-Bromo-4-nitrobenzoic acid
Formel:
C₇H₄BrNO₄ MW: 246.02 g/mol |
MDL Number:
MFCD00234252 CAS-Nummer: 16426-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...