4-Brombenzamidin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS1625G1 AS1625G5
ACTIAS1625G1EA 0 CHF
ACTIAS1625G1 ACTIAS1625G5
4-Brombenzamidin Hydrochlorid
4-Brombenzamidin Hydrochlorid
Formel: C₇H₈BrClN₂
MDL Number: MFCD00464967
CAS-Nummer: 55368-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR