3-Brom-4-hydroxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32322G10 AS32322G5
ACTIAS32322G10EA 0 CHF
ACTIAS32322G10 ACTIAS32322G5
3-Brom-4-hydroxybenzonitril
3-Brom-4-hydroxybenzonitril
Formel: C₇H₄BrNO
MW: 198.02 g/mol
Siedepunkt: 406
Melting Pt: 155
MDL Number: MFCD00143096
CAS-Nummer: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR