4-Brom-2-methoxybenzonitril
Lieferant: ACTIVATE SCIENTIFIC
AS30894G1
AS30894G5
ACTIAS30894G1EA
0
CHF
ACTIAS30894G1
ACTIAS30894G5
4-Brom-2-methoxybenzonitril
4-Brom-2-methoxybenzonitril
Formel:
C₈H₆BrNO |
MDL Number:
MFCD06738651 CAS-Nummer: 330793-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...