3-Brom-5-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32494G1 AS32494G5
ACTIAS32494G1EA 0 CHF
ACTIAS32494G1 ACTIAS32494G5
3-Brom-5-methoxybenzonitril
3-Brom-5-methoxybenzonitril
Formel: C₈H₆BrNO
MDL Number: MFCD08061971
CAS-Nummer: 867366-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR