2-Brom-5-chlorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS32254G1 AS32254G5
ACTIAS32254G1EA 0 CHF
ACTIAS32254G1 ACTIAS32254G5
2-Brom-5-chlorphenol
2-Brom-5-chlorphenol
Formel: C₆H₄BrClO
MW: 207.45 g/mol
MDL Number: MFCD00142870
CAS-Nummer: 13659-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR