5-Brom-3-chlor-2-methoxypyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methoxy-3-chloro-5-bromopyridine
AS33901G1
AS33901G10
AS33901G5
ACTIAS33901G1EA
0
CHF
ACTIAS33901G1
ACTIAS33901G10
ACTIAS33901G5
5-Brom-3-chlor-2-methoxypyridin
5-Brom-3-chlor-2-methoxypyridin
Formel:
C₆H₅BrClNO MW: 222,47 g/mol |
MDL Number:
MFCD04972382 CAS-Nummer: 848366-28-9 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...