3-Brom-5-chlor-2-methoxypyridin
Lieferant: ACTIVATE SCIENTIFIC
AS1860G1
AS1860G5
ACTIAS1860G1EA
0
CHF
ACTIAS1860G1
ACTIAS1860G5
3-Brom-5-chlor-2-methoxypyridin
3-Bromo-5-chloro-2-methoxypyridine
Formel:
C₆H₅BrClNO |
MDL Number:
MFCD08059322 CAS-Nummer: 102830-75-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...