3-Brom-4-chlor-5-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31815G1 AS31815G5
ACTIAS31815G1EA 0 CHF
ACTIAS31815G1 ACTIAS31815G5
3-Brom-4-chlor-5-nitropyridin
3-Bromo-4-chloro-5-nitropyridine
Formel: C₅H₂BrClN₂O₂
MDL Number: MFCD07368836
CAS-Nummer: 31872-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR