3-Brom-2-chlor-5-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31949G1 AS31949G5
ACTIAS31949G1EA 0 CHF
ACTIAS31949G1 ACTIAS31949G5
3-Brom-2-chlor-5-fluorpyridin
3-Bromo-2-chloro-5-fluoropyridine
Formel: C₅H₂BrClFN
MDL Number: MFCD03095269
CAS-Nummer: 884494-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR