5-Brom-2-chlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32416G1 AS32416G5
ACTIAS32416G1EA 0 CHF
ACTIAS32416G1 ACTIAS32416G5
5-Brom-2-chlorbenzonitril
5-Brom-2-chlorbenzonitril
Formel: C₇H₃BrClN
MW: 216,46 g/mol
MDL Number: MFCD00672948
CAS-Nummer: 57381-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR