2-Brom-4-chlorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS51603G1 AS51603G5
ACTIAS51603G1EA 0 CHF
ACTIAS51603G1 ACTIAS51603G5
2-Brom-4-chlorbenzonitril
2-Brom-4-chlorbenzonitril
Formel: C₇H₃BrClN
MW: 216,46 g/mol
MDL Number: MFCD00158966
CAS-Nummer: 57381-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR