3-Brom-2-chlor-6-methylpyridin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromo-2-chloro-6-picoline, 5-Bromo-6-chloro-2-picoline, 5-Bromo-6-chloro-2-methylpyridine
AS33730G1
AS33730G10
AS33730G5
ACTIAS33730G1EA
0
CHF
ACTIAS33730G1
ACTIAS33730G10
ACTIAS33730G5
3-Brom-2-chlor-6-methylpyridin
3-Bromo-2-chloro-6-methylpyridine
Formel:
C₆H₅BrClN MW: 206,47 g/mol |
MDL Number:
MFCD03095218 CAS-Nummer: 185017-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...