2-Brom-5-chlor-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-chloro-3-picoline

AS31708G1 AS31708G5
ACTIAS31708G1EA 0 CHF
ACTIAS31708G1 ACTIAS31708G5
2-Brom-5-chlor-3-methylpyridin
2-Bromo-5-chloro-3-methylpyridine
Formel: C₆H₅BrClN
MDL Number: MFCD03095062
CAS-Nummer: 65550-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR