3-Brom-6-chlorpyridazin
Lieferant: ACTIVATE SCIENTIFIC
AS30573G1
AS30573G5
ACTIAS30573G1EA
0
CHF
ACTIAS30573G1
ACTIAS30573G5
3-Brom-6-chlorpyridazin
3-Bromo-6-chloropyridazine
Formel:
C₄H₂BrClN₂ MW: 193.43 g/mol |
MDL Number:
MFCD09834814 CAS-Nummer: 89089-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...