4-Bromsalicylaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-2-hydroxybenzaldehyde

AS1670G1 AS1670G5
ACTIAS1670G1EA 0 CHF
ACTIAS1670G1 ACTIAS1670G5
4-Bromsalicylaldehyd
4-Bromsalicylaldehyd
Formel: C₇H₅BrO₂
MDL Number: MFCD06252606
CAS-Nummer: 22532-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR