2-Brom-3-hydroxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS34025G1 AS34025G10 AS34025G5
ACTIAS34025G1EA 0 CHF
ACTIAS34025G1 ACTIAS34025G10 ACTIAS34025G5
2-Brom-3-hydroxybenzaldehyd
2-Bromo-3-hydroxybenzaldehyde
Formel: C₇H₅BrO₂
MW: 201,02 g/mol
MDL Number: MFCD00092081
CAS-Nummer: 196081-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR