2-Brom-5-hydroxy-4-methoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31233G10 AS31233G5
ACTIAS31233G10EA 0 CHF
ACTIAS31233G10 ACTIAS31233G5
2-Brom-5-hydroxy-4-methoxybenzaldehyd
2-Bromo-5-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00195552
CAS-Nummer: 2973-59-3
EINECS: 426-540-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR