5-Brom-1-methylimidazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-1-methyl-1H-imidazole
AS50191G1
AS50191G5
ACTIAS50191G1EA
0
CHF
ACTIAS50191G1
ACTIAS50191G5
5-Brom-1-methylimidazol
5-Bromo-1-methylimidazole
Formel:
C₄H₅BrN₂ MW: 161 g/mol |
MDL Number:
MFCD01632218 CAS-Nummer: 1003-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...