2,5-Dibrom-4-methylimidazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,5-Dibromo-4-methyl-1H-imidazole

AS31793G1 AS31793G5
ACTIAS31793G1EA 0 CHF
ACTIAS31793G1 ACTIAS31793G5
2,5-Dibrom-4-methylimidazol
2,5-Dibromo-4-methylimidazole
Formel: C₄H₄Br₂N₂
MDL Number: MFCD02179519
CAS-Nummer: 219814-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR