4-Bromimidazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Bromo-1H-imidazole
AS50197G100
AS50197G5
ACTIAS50197G100EA
0
CHF
ACTIAS50197G100
ACTIAS50197G5
4-Bromimidazol
4-Bromoimidazole
Formel:
C₃H₃BrN₂ MW: 146,97 g/mol |
MDL Number:
MFCD00047021 CAS-Nummer: 2302-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...