2-Bromimidazol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Bromo-1H-imidazole
AS1789G1
AS1789G5
ACTIAS1789G1EA
31
CHF
ACTIAS1789G1
ACTIAS1789G5
2-Bromimidazol
2-Bromoimidazole
Formel:
C₃H₃BrN₂ MW: 146.97 g/mol |
MDL Number:
MFCD02179526 CAS-Nummer: 16681-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...