Furan-2-boronsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
Furan-2-ylboronic acid
AS50756G100
AS50756G5
ACTIAS50756G100EA
0
CHF
ACTIAS50756G100
ACTIAS50756G5
Furan-2-boronsäure
Furan-2-boronsäure
Formel:
C₄H₅BO₃ MW: 111,89 g/mol |
MDL Number:
MFCD00799544 CAS-Nummer: 13331-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...