1,2,3-Triazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole

AS22081G100 AS22081G25
ACTIAS22081G100EA 0 CHF
ACTIAS22081G100 ACTIAS22081G25
1,2,3-Triazol
1,2,3-Triazole
Formel: C₂H₃N₃
MW: 69,07 g/mol
Siedepunkt: 208 °C (1013 hPa)
Melting Pt: 23…25 °C
Dichte: 1,192 g/cm³ (20 °C)
MDL Number: MFCD00014490
CAS-Nummer: 288-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR