5-Fluor-2-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35079G1 AS35079G10 AS35079G5
ACTIAS35079G1EA 0 CHF
ACTIAS35079G1 ACTIAS35079G10 ACTIAS35079G5
5-Fluor-2-(trifluormethyl)benzonitril
5-Fluor-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
MDL Number: MFCD00236281
CAS-Nummer: 240800-45-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR