2-Fluor-6-(trifluormethyl)benzonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Cyano-3-fluorobenzotrifluoride
AS32251G1
AS32251G10
AS32251G5
ACTIAS32251G1EA
0
CHF
ACTIAS32251G1
ACTIAS32251G10
ACTIAS32251G5
2-Fluor-6-(trifluormethyl)benzonitril
2-Fluor-6-(trifluormethyl)benzonitril
Formel:
C₈H₃F₄N MW: 189.11 g/mol Siedepunkt: 212 Melting Pt: 26…28 °C Dichte: 1.373 g/cm³ (20 °C) |
MDL Number:
MFCD00061153 CAS-Nummer: 133116-83-3 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...