3-Chlor-5-(trifluormethyl)benzonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Chlor-5-cyanbenzotrifluorid, 3-Chlor-5-(trifluormethyl)benzoesäurenitril
AS35047G1
AS35047G10
AS35047G5
ACTIAS35047G1EA
0
CHF
ACTIAS35047G1
ACTIAS35047G10
ACTIAS35047G5
3-Chlor-5-(trifluormethyl)benzonitril
3-Chlor-5-(trifluormethyl)benzonitril
Formel:
C₈H₃ClF₃N MW: 205.57 |
MDL Number:
MFCD04115854 CAS-Nummer: 693245-52-2 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...