4-Brom-2,5-dimethoxytoluol

Lieferant: ACTIVATE SCIENTIFIC

AS35803G1 AS35803G10 AS35803G5
ACTIAS35803G1EA 0 CHF
ACTIAS35803G1 ACTIAS35803G10 ACTIAS35803G5
4-Brom-2,5-dimethoxytoluol
4-Brom-2,5-dimethoxytoluol
Formel: C₉H₁₁BrO₂
MW: 231,09 g/mol
MDL Number: MFCD00666999
CAS-Nummer: 13321-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR