5-Brom-2-thiophenboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-thienylboronic acid, 5-Bromothiophene-2-boronic acid, (5-Bromothiophen-2-yl)boronic acid

AS50232G1 AS50232G5
ACTIAS50232G1EA 0 CHF
ACTIAS50232G1 ACTIAS50232G5
5-Brom-2-thiophenboronsäure
5-Bromo-2-thiopheneboronic acid
Formel: C₄H₄BBrO₂S
MW: 206,85 g/mol
MDL Number: MFCD02094028
CAS-Nummer: 162607-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR