3-Thiophencarbonitril
Lieferant: ACTIVATE SCIENTIFIC
AS34461G10
AS34461G100
ACTIAS34461G10EA
0
CHF
ACTIAS34461G10
ACTIAS34461G100
3-Thiophencarbonitril
3-Thiophencarbonitril
Formel:
C₅H₃NS Siedepunkt: 82 °C (1013 hPa) Dichte: 1,1956 g/cm³ (20 °C) |
MDL Number:
MFCD00151852 CAS-Nummer: 1641-09-4 EINECS: 216-687-7 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...