3-Bromstyrol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-Bromo-3-vinylbenzene
AS24781G25
AS24781G5
ACTIAS24781G25EA
0
CHF
ACTIAS24781G25
ACTIAS24781G5
3-Bromstyrol
3-Bromstyrol
Formel:
C₈H₇Br MW: 183.05 g/mol Siedepunkt: 48.1…48.5 °C (0.5 mmHg) Dichte: 1.4059 |
MDL Number:
MFCD00000088 CAS-Nummer: 2039-86-3 EINECS: 218-025-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...