5-Nitro-o-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-5-nitrobenzonitrile

AS21391G100 AS21391G25
ACTIAS21391G100EA 248 CHF
ACTIAS21391G100 ACTIAS21391G25
5-Nitro-o-toluonitril
5-Nitro-o-toluonitril
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Siedepunkt: 174 °C (18 mmHg)
Melting Pt: 104…106 °C
MDL Number: MFCD00017040
CAS-Nummer: 939-83-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR