2,4,6-Trimethylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS25274G100 AS25274G25
ACTIAS25274G100EA 0 CHF
ACTIAS25274G100 ACTIAS25274G25
2,4,6-Trimethylbenzonitril
2,4,6-Trimethylbenzonitril
Formel: C₁₀H₁₁N
MW: 145.2 g/mol
MDL Number: MFCD00016378
CAS-Nummer: 2571-52-0
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR