3-Cyan-6-fluoranilin
Lieferant: ACTIVATE SCIENTIFIC
AS32491G1
AS32491G5
ACTIAS32491G1EA
0
CHF
ACTIAS32491G1
ACTIAS32491G5
3-Cyan-6-fluoranilin
3-Cyan-6-fluoranilin
Formel:
C₇H₅FN₂ |
MDL Number:
MFCD07787428 CAS-Nummer: 859855-53-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...