3-Cyan-5-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-5-fluorobenzonitrile, 3-Cyano-5-fluoroaniline

AS35468G1 AS35468G5
ACTIAS35468G1EA 0 CHF
ACTIAS35468G1 ACTIAS35468G5
3-Cyan-5-fluoranilin
3-Cyan-5-fluoranilin
Formel: C₇H₅FN₂
MW: 136,13 g/mol
MDL Number: MFCD05864357
CAS-Nummer: 210992-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR