2-Cyan-5-methylanilin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Amino-4-methylbenzonitril, 2-Amino-4-methylbenzoesäurenitril
AS32332G1
AS32332G10
AS32332G5
ACTIAS32332G1EA
0
CHF
ACTIAS32332G1
ACTIAS32332G10
ACTIAS32332G5
2-Cyan-5-methylanilin
2-Cyan-5-methylanilin
Formel:
C₈H₈N₂ |
MDL Number:
MFCD00173706 CAS-Nummer: 26830-96-6 EINECS: 248-020-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...