D-α-Phenylalanin

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Phenyl-D(+)-alanin3-Phenyl-D-alaninH-D-Phe-OH(R)-(+)-2-Amino-3-phenylpropionsäure(R)-(+)-2-Amino-3-phenylpropansäure(R)-(+)-2-Amino-3-phenylpropionic acidD-Phenylalanin

ACTIAS21706G25EA 45 CHF
ACTIAS21706G25 ACTIAS21706G100
D-α-Phenylalanin
D-α-Phenylalanin
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
Siedepunkt: 295 °C (1013 hPa)
MDL Number: MFCD00004270
CAS-Nummer: 673-06-3
EINECS: 211-603-5
Merck Index: 12,07425

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR