2-Fluor-5-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid
AS50618G1
AS50618G5
ACTIAS50618G1EA
0
CHF
ACTIAS50618G1
ACTIAS50618G5
2-Fluor-5-pyridinboronsäure
2-Fluor-5-pyridinboronsäure
Formel:
C₅H₅BFNO₂ |
MDL Number:
MFCD03411559 CAS-Nummer: 351019-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...