2-Brom-5-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid
AS50033G1
AS50033G5
ACTIAS50033G1EA
0
CHF
ACTIAS50033G1
ACTIAS50033G5
2-Brom-5-pyridinboronsäure
2-Brom-5-pyridinboronsäure
Formel:
C₅H₅BBrNO₂ MW: 201.81 g/mol |
MDL Number:
MFCD03411558 CAS-Nummer: 223463-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...