2-Fluor-4-pyridinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid
AS50169G1
AS50169G5
ACTIAS50169G1EA
0
CHF
ACTIAS50169G1
ACTIAS50169G5
2-Fluor-4-pyridinboronsäure
2-Fluoro-4-pyridineboronic acid
Formel:
C₅H₅BFNO₂ MW: 140.91 g/mol |
MDL Number:
MFCD04112534 CAS-Nummer: 401815-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...