2-Chlor-4-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51138G1 AS51138G5
ACTIAS51138G1EA 18 CHF
ACTIAS51138G1 ACTIAS51138G5
2-Chlor-4-pyridinboronsäure
2-Chloro-4-pyridineboronic acid
Formel: C₅H₅BClNO₂
MDL Number: MFCD03788416
CAS-Nummer: 458532-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR