2-Chlor-3-pyridinboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3-pyridinylboronic acid, (2-Chloropyridin-3-yl)boronic acid, 2-Chloropyridine-3-boronic acid

AS50722G1 AS50722G5
ACTIAS50722G1EA 0 CHF
ACTIAS50722G1 ACTIAS50722G5
2-Chlor-3-pyridinboronsäure
2-Chloro-3-pyridineboronic acid
Formel: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03094997
CAS-Nummer: 381248-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR