2-Cyan-4-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS21297G25 AS21297G5
ACTIAS21297G25EA 206 CHF
ACTIAS21297G25 ACTIAS21297G5
2-Cyan-4-fluoranilin
2-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
MW: 136,13 g/mol
MDL Number: MFCD02683869
CAS-Nummer: 61272-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR