4-Methylnicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Methylpyridine-3-carboxylic acid, 4-Methyl-3-pyridinecarboxylic acid

AS7061G1 AS7061G5
ACTIAS7061G1EA 0 CHF
ACTIAS7061G1 ACTIAS7061G5
4-Methylnicotinsäure
4-Methylnicotinsäure
Formel: C₇H₇NO₂
MW: 137,14 g/mol
MDL Number: MFCD00128869
CAS-Nummer: 3222-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR