4-Chlornicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-3-pyridinecarboxylic acid, 4-Chloropyridine-3-carboxylic acid

AS22446G25 AS22446G5
ACTIAS22446G25EA 129 CHF
ACTIAS22446G25 ACTIAS22446G5
4-Chlornicotinsäure
4-Chlornicotinsäure
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
MDL Number: MFCD00128860
CAS-Nummer: 10177-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR