4-Chlor-3-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS50137G1 AS50137G100 AS50137G5
ACTIAS50137G1EA 0 CHF
ACTIAS50137G1 ACTIAS50137G100 ACTIAS50137G5
4-Chlor-3-fluorphenylboronsäure
4-Chlor-3-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
MDL Number: MFCD01319010
CAS-Nummer: 137504-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR