4-Bromthiazol

Lieferant: ACTIVATE SCIENTIFIC

AS50106G1 AS50106G5
ACTIAS50106G1EA 0 CHF
ACTIAS50106G1 ACTIAS50106G5
4-Bromthiazol
4-Bromothiazole
Formel: C₃H₂BrNS
MW: 164,03 g/mol
MDL Number: MFCD06657592
CAS-Nummer: 34259-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR